Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141931.1 | NP_001135403.1 | 12 | 1333 | ||
NM_001141932.1 | NP_001135404.1 | 13 | 1381 | |||
NM_020296.2 | NP_064692.2 | 12 | 1526 | |||
GenBank mRNA | AB026569.1 | - | 12 | 1249 | ||
AK133054.1 | - | 12 | 1054 | |||
AK135280.1 | - | 12 | 1213 | |||
AK145897.1 | - | 12 | 1335 | |||
AK147109.1 | - | 12 | 1527 | |||
AK161102.1 | - | 13 | 1331 | |||
AK161416.1 | - | 8 | 663 | |||
AK203574.1 | - | 4 | 280 | |||
BC016501.1 | - | 12 | 1514 | 4527742 | ||
BC056609.1 | - | 10 | 773 | |||
BC057866.1 | - | 13 | 1413 | 5327143 |