Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd1lg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021396.2 | NP_067371.1 | 3 | 575 | ||
GenBank mRNA | AF142780.2 | - | 3 | 548 | ||
AK089369.1 | - | 3 | 578 | |||
AK141111.1 | - | 2 | 665 | |||
AK155002.1 | - | 3 | 515 | |||
BC104138.1 | - | 3 | 480 | 40045578 | ||
BC104139.1 | - | 3 | 480 | 40045580 | ||
BY749628.1 | - | 3 | 578 |