Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253832.1 | NP_001240761.1 | 8 | 979 | ||
NM_021438.4 | NP_067413.1 | 8 | 958 | |||
GenBank mRNA | AF270700.1 | - | 8 | 871 | ||
AK008093.1 | - | 6 | 1301 | |||
AK008132.1 | - | 8 | 962 | |||
AK012629.1 | - | 8 | 941 | |||
AK019399.1 | - | 8 | 950 | |||
AK028029.1 | - | 8 | 949 | |||
AK086610.1 | - | 3 | 877 | |||
BC031716.1 | - | 8 | 949 | 3969355 | ||
BC049284.1 | - | 8 | 957 | 6395407 |