Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162858.1 | NP_001156330.1 | 2, 3 | 686 | ||
NM_021717.2 | NP_068363.2 | 2, 3 | 686 | |||
GenBank mRNA | AJ278170.1 | - | 2, 3 | 709 | ||
AK134689.1 | - | 2, 3 | 483 | |||
AK156526.1 | - | 2, 3 | 559 | |||
BC115523.1 | - | 2, 3 | 436 | 40101227 | ||
BY718516.1 | - | 2, 3 | 481 |