Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022884.2 | NP_075022.2 | 2 | 168 | ||
GenBank mRNA | AF257474.1 | - | 2 | 154 | ||
AK090308.1 | - | 2 | 146 | |||
AK145901.1 | - | 2 | 169 | |||
BC013515.1 | - | 2 | 165 | 4235455 |