Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025316.2 | NP_079592.2 | 4, 5 | 350 | ||
GenBank mRNA | AK002279.1 | - | 4, 5 | 352 | ||
AK004372.1 | - | 4, 5 | 350 | |||
AK005521.1 | - | 4, 5 | 350 | |||
AK013223.1 | - | 4, 5 | 352 | |||
AK018562.1 | - | 4, 5 | 357 | |||
AK088061.1 | - | 4, 5 | 359 | |||
BC025155.1 | - | 4, 5 | 343 | 4024605 |