Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160145.1 | NP_001153617.1 | 2, 3 | 297 | ||
NM_001160146.1 | NP_001153618.1 | 2, 3 | 193 | |||
NM_025439.3 | NP_079715.1 | 2, 3 | 162 | |||
GenBank mRNA | AK005248.1 | - | 2, 3 | 149 | ||
AK021225.1 | - | 2, 3 | 149 | |||
AK134909.1 | - | 2, 3 | 127 | |||
AK154423.1 | - | 2, 3 | 149 | |||
AK190183.1 | - | 1, 2 | 107 | |||
BC016524.1 | - | 2, 3 | 137 | 3594475 | ||
BC020291.1 | - | 1, 2 | 114 | |||
BC048017.1 | - | 2, 3 | 116 | |||
BQ918002.1 | - | 2, 3 | 384 | |||
CA460035.1 | - | 2, 3 | 199 | |||
CX208138.1 | - | 2, 3 | 193 |