Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025473.3 | NP_079749.1 | 6, 7 | 847 | ||
GenBank mRNA | AK007715.1 | - | 6, 7 | 848 | ||
AK012353.1 | - | 2, 3 | 175 | |||
BC011237.1 | - | 6, 7 | 845 | |||
BC019752.1 | - | 7, 8 | 613 | |||
BC054411.1 | - | 6, 7 | 519 | |||
BC069871.1 | - | 6, 7 | 982 | 6467541 |