Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025544.2 | NP_079820.2 | 7 | 664 | ||
GenBank mRNA | AB049947.1 | - | 7 | 540 | ||
AK003126.1 | - | 7 | 585 | |||
AK008623.1 | - | 7 | 616 | |||
AK010330.1 | - | 7 | 627 | |||
AK050396.1 | - | 7 | 668 | |||
BC029193.1 | - | 7 | 583 | 4457237 | ||
BC055861.1 | - | 7 | 563 | 3587953 | ||
CF582282.1 | - | 2 | 293 |