Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025544.2 | NP_079820.2 | 6, 7 | 590 | ||
GenBank mRNA | AB049947.1 | - | 6, 7 | 466 | ||
AK003126.1 | - | 6, 7 | 511 | |||
AK008623.1 | - | 6, 7 | 542 | |||
AK010330.1 | - | 6, 7 | 553 | |||
AK050396.1 | - | 6, 7 | 594 | |||
BC029193.1 | - | 6, 7 | 509 | 4457237 | ||
BC055861.1 | - | 6, 7 | 489 | 3587953 | ||
CF582282.1 | - | 2, 3 | 367 |
Gene Symbol: Ptov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK178502.1 | - | 37 |