Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190322.2 | NP_001177251.1 | 4 | 470 | ||
NM_001190323.2 | NP_001177252.1 | 4 | 346 | |||
NM_001190324.2 | NP_001177253.1 | 4 | 301 | |||
NM_001285804.1 | NP_001272733.1 | 4 | 501 | |||
NM_181391.4 | NP_852056.1 | 4 | 515 | |||
GenBank mRNA | AA980189.1 | - | 4 | 298 | ||
AI851035.1 | - | 2 | 192 | |||
AK003308.1 | - | 4 | 347 | |||
AK007844.1 | - | 4 | 490 | |||
AK013200.1 | - | 4 | 488 | |||
AK145105.1 | - | 4 | 490 | |||
AK168647.1 | - | 4 | 445 | |||
AK169018.1 | - | 4 | 490 | |||
BC030366.1 | - | 4 | 473 | 5370748 | ||
BX527959.1 | - | 4 | 262 | |||
CB057699.1 | - | 4 | 427 |