Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190322.2 | NP_001177251.1 | 3, 4 | 412 | ||
NM_001190323.2 | NP_001177252.1 | 3, 4 | 288 | |||
NM_001190324.2 | NP_001177253.1 | 3, 4 | 243 | |||
NM_001285804.1 | NP_001272733.1 | 3, 4 | 443 | |||
NM_181391.4 | NP_852056.1 | 3, 4 | 457 | |||
GenBank mRNA | AA980189.1 | - | 3, 4 | 240 | ||
AI851035.1 | - | 2, 3 | 250 | |||
AK003308.1 | - | 3, 4 | 289 | |||
AK007844.1 | - | 3, 4 | 432 | |||
AK013200.1 | - | 3, 4 | 430 | |||
AK145105.1 | - | 3, 4 | 432 | |||
AK168647.1 | - | 3, 4 | 387 | |||
AK169018.1 | - | 3, 4 | 432 | |||
BC030366.1 | - | 3, 4 | 415 | 5370748 | ||
BX527959.1 | - | 3, 4 | 204 | |||
CB057699.1 | - | 3, 4 | 369 |