Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181392.3 | NP_852057.2 | 3 | 176 | ||
GenBank mRNA | AK003547.1 | - | 3 | 165 | ||
AK003579.1 | - | 3 | 160 | |||
AK011281.1 | - | 3 | 161 | |||
AK012257.1 | - | 3 | 238 | |||
AK013225.1 | - | 3 | 140 | |||
AK019011.1 | - | 3 | 131 | |||
BC029238.1 | - | 3 | 152 | 4489531 | ||
W50582.1 | - | 3 | 175 |