Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181392.3 | NP_852057.2 | 3 | 247 | ||
GenBank mRNA | AK003547.1 | - | 3 | 236 | ||
AK003579.1 | - | 3 | 231 | |||
AK011281.1 | - | 3 | 232 | |||
AK012257.1 | - | 3 | 309 | |||
AK013225.1 | - | 3 | 211 | |||
AK019011.1 | - | 3 | 202 | |||
BC029238.1 | - | 3 | 223 | 4489531 | ||
W50582.1 | - | 3 | 246 |