Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025631.3 | NP_079907.2 | 14 | 1331 | ||
GenBank mRNA | AK009346.1 | - | 14 | 1303 | ||
AK009504.1 | - | 14 | 1306 | |||
AK009619.1 | - | 14 | 1304 | |||
AK009754.1 | - | 10 | 907 | |||
AK009776.1 | - | 14 | 1289 | |||
AK009794.1 | - | 14 | 1291 | |||
AK009871.1 | - | 14 | 1300 | |||
AK010130.1 | - | 14 | 1286 | |||
AK010154.1 | - | 14 | 1299 | |||
AK010157.1 | - | 14 | 1330 | |||
AK010215.1 | - | 14 | 1300 | |||
AK028156.1 | - | 14 | 1302 | |||
AK028158.1 | - | 14 | 1305 | |||
AK160516.1 | - | 14 | 1302 | |||
BC116681.1 | - | 14 | 1286 | 40106489 | ||
BC116727.1 | - | 14 | 1286 | 40106498 |