Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025901.3 | NP_080177.1 | 2 | 224 | ||
GenBank mRNA | AK005152.1 | - | 2 | 128 | ||
AK008637.1 | - | 2 | 96 | |||
AK010875.1 | - | 2 | 162 | |||
AK021353.1 | - | 2 | 159 | |||
AK031110.1 | - | 2 | 159 | |||
AK088206.1 | - | 2 | 124 | |||
BC025898.1 | - | 2 | 240 | |||
BY003788.1 | - | 2 | 226 |