Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025999.3 | NP_080275.2 | 5, 6 | 639 | ||
GenBank mRNA | AF353167.1 | - | 5, 6 | 669 | ||
AK006791.1 | - | 5, 6 | 679 | |||
AK011839.1 | - | 5, 6 | 628 | |||
AK015505.1 | - | 5, 6 | 640 | |||
AK139910.1 | - | 5, 6 | 633 | |||
AK154417.1 | - | 5, 6 | 663 | |||
BC018553.1 | - | 5, 6 | 633 | 3582546 |