Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026025.2 | NP_080301.1 | 3, 4 | 232 | ||
GenBank mRNA | AB095989.1 | - | 3, 4 | 265 | ||
AK012852.1 | - | 3, 4 | 233 | |||
AK170136.1 | - | 3, 4 | 244 | |||
BC058368.1 | - | 3, 4 | 266 | 6819507 |