Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026027.3 | NP_080303.1 | 2 | 179 | ||
GenBank mRNA | AK002575.1 | - | 2 | 178 | ||
AK003330.1 | - | 2 | 197 | |||
AK010532.1 | - | 2 | 193 | |||
AK012575.1 | - | 2 | 180 | |||
AK028148.1 | - | 2 | 185 | |||
BC024693.1 | - | 2 | 202 | 4020579 | ||
BC094627.1 | - | 2 | 180 | 1532309 |