Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164237.1 | NP_001157709.1 | 7 | 1204 | ||
NM_026259.3 | NP_080535.2 | 7 | 1204 | |||
GenBank mRNA | AF305730.1 | - | 7 | 1165 | ||
AK015745.1 | - | 6 | 924 | |||
AK016825.1 | - | 6 | 784 | |||
AK020527.1 | - | 5 | 777 | |||
AK028551.1 | - | 7 | 1075 | |||
AK030558.1 | - | 7 | 1203 | |||
AK031657.1 | - | 6 | 846 | |||
AK032848.1 | - | 7 | 1107 | |||
AK034551.1 | - | 7 | 1120 | |||
AK044103.1 | - | 6 | 836 | |||
AK045760.1 | - | 7 | 1188 | |||
AK077466.1 | - | 7 | 1063 | |||
BC019415.1 | - | 4 | 573 | 4014525 | ||
BC049078.1 | - | 7 | 1163 | 6419123 |