Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164237.1 | NP_001157709.1 | 7 | 1073 | ||
NM_026259.3 | NP_080535.2 | 7 | 1073 | |||
GenBank mRNA | AF305730.1 | - | 7 | 1034 | ||
AK015745.1 | - | 6 | 793 | |||
AK016825.1 | - | 6 | 653 | |||
AK020527.1 | - | 5 | 646 | |||
AK028551.1 | - | 7 | 944 | |||
AK030558.1 | - | 7 | 1072 | |||
AK031657.1 | - | 6 | 715 | |||
AK032848.1 | - | 7 | 976 | |||
AK034551.1 | - | 7 | 989 | |||
AK044103.1 | - | 6 | 705 | |||
AK045760.1 | - | 7 | 1057 | |||
AK077466.1 | - | 7 | 932 | |||
BC019415.1 | - | 4 | 442 | 4014525 | ||
BC049078.1 | - | 7 | 1032 | 6419123 |