Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026307.3 | NP_080583.3 | 7 | 597 | ||
NM_026948.3 | NP_081224.1 | 6 | 545 | |||
GenBank mRNA | AK002828.1 | - | 7 | 541 | ||
AK003381.1 | - | 6 | 485 | |||
AK003647.1 | - | 7 | 539 | |||
AK004197.1 | - | 7 | 539 | |||
AK007580.1 | - | 5 | 674 | |||
AK012608.1 | - | 4 | 1017 | |||
AK159336.1 | - | 7 | 544 | |||
AK199328.1 | - | 4 | 204 | |||
AK207668.1 | - | 3 | 279 | |||
BC021463.1 | - | 5 | 659 | |||
BC024422.1 | - | 7 | 527 | 5005082 | ||
BC028531.1 | - | 6 | 636 | |||
BC037500.1 | - | 4 | 627 | |||
CA323205.1 | - | 6 | 510 | |||
CK624897.1 | - | 6 | 522 |