Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026316.2 | NP_080592.2 | 4 | 343 | ||
GenBank mRNA | AK154500.1 | - | 4 | 389 | ||
AK154723.1 | - | 4 | 436 | |||
AK159447.1 | - | 4 | 357 | |||
AK160106.1 | - | 4 | 360 | |||
BC046597.1 | - | 4 | 343 | 6312211 |