Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110004E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026502.2 | NP_080778.2 | 4 | 577 | ||
GenBank mRNA | AK003413.1 | - | 4 | 468 | ||
AK018510.1 | - | 4 | 548 | |||
AK045905.1 | - | 4 | 556 | |||
BC019533.1 | - | 4 | 541 | 4191729 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK028807.1 | - | 1915 | Chr. - - [-] on Build |