Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp85os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015755.1 | - | 3 | 564 | ||
AK019608.1 | - | 3 | 613 | |||
AK134579.1 | - | 3 | 897 | |||
BC145678.1 | - | 1 | 157 |