Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085409.1 | NP_001078878.1 | 5 | 1578 | ||
NM_133186.3 | NP_573449.2 | 4 | 1205 | |||
GenBank mRNA | AK003108.1 | - | 1179 | |||
AK028961.1 | - | 4 | 1182 | |||
AK078769.1 | - | 4 | 1599 | |||
AK167028.1 | - | 5 | 1578 | |||
AK171237.1 | - | 4 | 1210 | |||
AY029586.1 | - | 4 | 1193 | |||
BC027251.1 | - | 4 | 1188 | |||
BC037435.1 | - | 4 | 1206 | 2651584 |