Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183275.2 | NP_899098.2 | 4 | 862 | ||
GenBank mRNA | AK003325.1 | - | 4 | 839 | ||
AK146639.1 | - | 4 | 863 | |||
BC019666.1 | - | 1 | 176 | |||
BC094625.1 | - | 2 | 296 | 1515023 | ||
BC132029.1 | - | 3 | 502 |