Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ict1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026729.1 | NP_081005.1 | 5 | 334 | ||
GenBank mRNA | AK003192.1 | - | 5 | 334 | ||
AK003286.1 | - | 5 | 306 | |||
AK160697.1 | - | 123 | ||||
BC028523.1 | - | 5 | 435 | 2938122 | ||
BC079614.1 | - | 373 |