Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hddc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026812.2 | NP_081088.1 | 4 | 426 | ||
GenBank mRNA | AK004078.1 | - | 4 | 427 | ||
AK031866.1 | - | 2327 | ||||
AK080642.1 | - | 225 | ||||
BC038310.1 | - | 4 | 439 | 5119020 | ||
BC055847.1 | - | 3 | 1035 |