Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254762.1 | NP_001241691.1 | 2 | 487 | ||
NM_030037.2 | NP_084313.1 | 3 | 640 | |||
GenBank mRNA | AK088731.1 | - | 2 | 478 | ||
BC003880.1 | - | 2 | 497 | 3496491 | ||
BC034167.1 | - | 3 | 625 | |||
BC039620.1 | - | 3 | 621 | 5051528 | ||
BE861793.1 | - | 2 | 114 | |||
BQ445267.1 | - | 1 | 53 | |||
CK617131.1 | - | 2 | 438 |