Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026902.3 | NP_081178.1 | 6 | 637 | ||
GenBank mRNA | AK005292.1 | - | 6 | 638 | ||
AK087975.1 | - | 6 | 641 | |||
AK138385.1 | - | 6 | 656 | |||
BC010486.1 | - | 6 | 610 | 3483367 |