Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027213.2 | NP_081489.2 | 8 | 722 | ||
GenBank mRNA | AK005232.1 | - | 8 | 768 | ||
AK171784.1 | - | 8 | 739 | |||
BC013096.1 | - | 8 | 722 | 3710612 |