Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027213.2 | NP_081489.2 | 5 | 398 | ||
GenBank mRNA | AK005232.1 | - | 5 | 443 | ||
AK037414.1 | - | 1 | 1496 | |||
AK089101.1 | - | 5 | 447 | |||
AK171784.1 | - | 5 | 414 | |||
BC013096.1 | - | 5 | 398 | 3710612 |