Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1fnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027304.2 | NP_081580.2 | 1 | 1166 | ||
GenBank mRNA | AB016273.1 | - | 1 | 1138 | ||
AK006402.1 | - | 1 | 904 | |||
AY496853.1 | - | 1 | 1164 | |||
BC052363.1 | - | 1 | 1152 | 6773647 |