Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011942.1 | - | 3 | 750 | ||
AK053363.1 | - | 3 | 709 | |||
AK150539.1 | - | 3 | 868 | |||
AK151163.1 | - | 3 | 868 | |||
AK154611.1 | - | 3 | 819 | |||
BC020064.1 | - | 3 | 774 | |||
BC048708.1 | - | 3 | 642 | |||
BC060191.1 | - | 3 | 743 | |||
BC063765.1 | - | 3 | 685 | |||
BC080823.1 | - | 3 | 695 | |||
BC116944.1 | - | 3 | 793 | 40126263 | ||
BC116970.1 | - | 3 | 793 | 40126289 | ||
BC144746.1 | - | 3 | 693 |