Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027866.1 | NP_082142.1 | 7 | 856 | ||
GenBank mRNA | AK003121.1 | - | 7 | 856 | ||
BC104380.1 | - | 7 | 827 | 40050136 | ||
BC104381.1 | - | 7 | 845 | 40050143 |