Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027941.1 | NP_082217.1 | 9 | 987 | ||
GenBank mRNA | AK005720.1 | - | 9 | 1039 | ||
BC100519.1 | - | 9 | 987 | 30919449 | ||
BC116419.1 | - | 8 | 820 | |||
BC116420.1 | - | 8 | 827 | 40103399 | ||
JF934738.1 | - | 9 | 938 |