Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164827.1 | NP_001158299.1 | 2, 3 | 588 | ||
NM_028054.3 | NP_082330.2 | 2, 3 | 588 | |||
GenBank mRNA | AK005386.1 | - | 2, 3 | 352 | ||
BC099429.1 | - | 2, 3 | 281 | 30790629 |