Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028055.4 | NP_082331.1 | 3 | 479 | ||
GenBank mRNA | AK005160.1 | - | 3 | 485 | ||
AK032100.1 | - | 3 | 481 | |||
AK046050.1 | - | 3 | 480 | |||
BC057599.1 | - | 2 | 1756 | |||
BC060053.1 | - | 2 | 1787 | |||
BC122876.1 | - | 3 | 462 | 40107633 |