Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031373.3 | NP_113550.3 | 1, 2 | 211 | ||
GenBank mRNA | AF303894.1 | - | 1, 2 | 159 | ||
AK008230.1 | - | 1, 2 | 176 | |||
AK149146.1 | - | 1, 2 | 158 | |||
AK150115.1 | - | 1, 2 | 144 | |||
BC003875.1 | - | 1, 2 | 142 | 3495186 | ||
BC049965.1 | - | 1, 2 | 157 | 3155091 | ||
CJ144579.1 | - | 1, 2 | 209 |