Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176835.2 | NP_789805.1 | 2 | 686 | ||
GenBank mRNA | AK013323.1 | - | 2 | 709 | ||
AK050294.1 | - | 2 | 665 | |||
BC039633.1 | - | 2 | 686 | 5136672 |