Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028417.1 | NP_082693.1 | 2 | 474 | ||
GenBank mRNA | AK013779.1 | - | 2 | 477 | ||
BC048750.1 | - | 2 | 472 | |||
BC089566.1 | - | 2 | 474 | 6531894 |