Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204241.1 | NP_001191170.1 | 3, 4 | 285 | ||
NM_153197.4 | NP_694877.2 | 2, 3 | 421 | |||
GenBank mRNA | AK014135.1 | - | 2, 3 | 321 | ||
AK156040.1 | - | 2, 3 | 421 | |||
AY397674.1 | - | 2, 3 | 232 | |||
BC034893.1 | - | 3, 4 | 285 | 1496162 |