Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcee | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028626.1 | NP_082902.1 | 2 | 93 | ||
GenBank mRNA | AK003511.1 | - | 2 | 90 | ||
BC038157.1 | - | 2 | 93 | 1479934 |