Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019715.2 | NP_062689.2 | 5 | 898 | ||
GenBank mRNA | AK007070.1 | - | 5 | 842 | ||
AK142436.1 | - | 5 | 655 | |||
BC029325.1 | - | 5 | 720 | |||
BC043330.1 | - | 5 | 679 | 5249829 |