Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028881.2 | NP_083157.1 | 13 | 1961 | ||
GenBank mRNA | AA276938.1 | - | 2 | 186 | ||
AK014553.1 | - | 7 | 2068 | |||
AK079382.1 | - | 10 | 2302 | |||
AK156192.1 | - | 13 | 1940 | |||
AK169944.1 | - | 9 | 1610 | |||
BC019713.1 | - | 3 | 343 | |||
BC023091.1 | - | 9 | 1551 | 4023064 | ||
BC023831.1 | - | 13 | 1956 | 5345301 | ||
BC032183.1 | - | 13 | 1966 | 5310152 | ||
BC033300.1 | - | 13 | 1956 | 4166411 | ||
CO040040.1 | - | 2 | 681 |