Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028959.3 | NP_083235.3 | 7, 8 | 1226 | ||
GenBank mRNA | AK017134.1 | - | 7, 8 | 1218 | ||
AK034892.1 | - | 7, 8 | 1219 | |||
AK161033.1 | - | 3, 4 | 551 | |||
BC100549.1 | - | 6, 7 | 1076 |