Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029512.2 | NP_083788.2 | 2 | 487 | ||
NM_181734.3 | NP_859423.2 | 2 | 484 | |||
GenBank mRNA | AK018052.1 | - | 2 | 448 | ||
AK033600.1 | - | 2 | 433 | |||
AK039450.1 | - | 2 | 485 | |||
AK043791.1 | - | 2 | 454 | |||
AK052794.1 | - | 2 | 450 | |||
AK081680.1 | - | 2 | 447 | |||
AK156641.1 | - | 2 | 450 | |||
BC052389.1 | - | 2 | 443 | 5718874 | ||
BC060726.1 | - | 1 | 353 | |||
BY717985.1 | - | 2 | 448 | |||
CK640285.1 | - | 1 | 120 |