Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164363.1 | NP_001157835.1 | 17 | 1908 | ||
NM_001164365.1 | NP_001157837.1 | 17 | 1565 | |||
NM_029810.4 | NP_084086.3 | 17 | 1496 | |||
GenBank mRNA | AK008045.1 | - | 17 | 1443 | ||
AK030786.1 | - | 18 | 1515 | |||
AK036730.1 | - | 17 | 1454 | |||
AK038458.1 | - | 14 | 1202 | |||
AK132384.1 | - | 17 | 1454 | |||
AK147901.1 | - | 17 | 1454 | |||
AK162385.1 | - | 17 | 1873 | |||
AK169616.1 | - | 17 | 1454 | |||
AK211993.1 | - | 1 | 13 | |||
BC064760.1 | - | 16 | 1430 | 30436992 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC006028.1 | - | 93 | Chr. - - [-] on Build |