Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029832.2 | NP_084108.1 | 3 | 446 | ||
GenBank mRNA | AK011170.1 | - | 3 | 377 | ||
BC051541.1 | - | 1 | 35 | |||
BC115616.2 | - | 3 | 446 | 40102754 | ||
BC115617.1 | - | 3 | 446 | 40102757 |